Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 4 | 497 | ||
NM_004901.4 | NP_004892.1 | 4 | 497 | |||
GenBank mRNA | AB002390.1 | - | 2 | 51 | ||
AF016032.1 | - | 4 | 426 | |||
AJ131358.1 | - | 4 | 394 | |||
AK300464.1 | - | 4 | 405 | |||
BC034477.2 | - | 4 | 461 | 4824559 | ||
CN313716.1 | - | 2 | 53 | |||
DC341200.1 | - | 4 | 497 |