Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138706.4 | NP_619651.3 | 2 | 1572 | ||
GenBank mRNA | AK127544.1 | - | 2 | 1155 | ||
AK172863.1 | - | 2 | 1497 | |||
BC025357.1 | - | 1 | 1113 | |||
BC103908.1 | - | 2 | 1471 | 40005856 | ||
BC103909.2 | - | 2 | 1471 | 40005863 | ||
BC103910.1 | - | 2 | 1471 | 40005864 |