Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014298.4 | NP_055113.2 | 2 | 711 | ||
XM_005255223.3 | XP_005255280.2 | 3 | 794 | |||
XM_017023101.1 | XP_016878590.1 | 3 | 794 | |||
GenBank mRNA | BC005060.1 | - | 2 | 548 | 2960170 | |
BC010033.2 | - | 2 | 540 | 3506460 | ||
BC018910.2 | - | 2 | 565 | 3959973 | ||
BT007231.1 | - | 1 | 521 | |||
EU794638.1 | - | 2 | 540 | |||
KJ534923.1 | - | 1 | 521 |