Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 2 | 469 | ||
NM_001256743.1 | NP_001243672.1 | 2 | 469 | |||
NM_007325.4 | NP_015564.4 | 2 | 469 | |||
GenBank mRNA | AK297136.1 | - | 2 | 155 | ||
BC032004.1 | - | 2 | 413 | |||
BC117464.1 | - | 2 | 341 | 40126015 | ||
DA191279.1 | - | 2 | 469 | |||
U10301.1 | - | 2 | 229 | |||
U10302.1 | - | 2 | 229 |