Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033557.2 | NP_001028729.1 | 3, 4 | 1458 | ||
NM_002706.5 | NP_002697.1 | 3, 4 | 1458 | |||
NM_177968.3 | NP_808907.1 | 3, 4 | 1458 | |||
NM_177969.3 | NP_808908.1 | 2, 3 | 598 | |||
XM_011532936.2 | XP_011531238.1 | 3, 4 | 1416 | |||
GenBank mRNA | AF136972.1 | - | 2, 3 | 972 | ||
AF294792.1 | - | 2, 3 | 972 | |||
AJ005801.1 | - | 2, 3 | 972 | |||
AJ271832.1 | - | 3, 4 | 1365 | |||
AJ271835.1 | - | 3, 4 | 1365 | |||
AK056009.1 | - | 3, 4 | 1394 | |||
AK291214.1 | - | 3, 4 | 1400 | |||
AL833035.1 | - | 3, 4 | 1263 | |||
BC012002.1 | - | 2, 3 | 500 | |||
BC064381.1 | - | 3, 4 | 1373 | 5734287 | ||
DQ023508.1 | - | 3, 4 | 1384 | |||
DQ023510.1 | - | 3, 4 | 1384 | |||
DQ023511.1 | - | 3, 4 | 1384 |