Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010578.2 | NP_034708.1 | 7, 8 | 1246 | ||
GenBank mRNA | AK076526.1 | - | 3, 4 | 543 | ||
AK088016.1 | - | 7, 8 | 1147 | |||
AK088729.1 | - | 7, 8 | 993 | |||
AK135521.1 | - | 6, 7 | 922 | |||
AK141253.1 | - | 2, 3 | 173 | |||
AK159689.1 | - | 7, 8 | 1088 | |||
AK167682.1 | - | 7, 8 | 1144 | |||
BC050906.1 | - | 7, 8 | 1119 | 5721348 | ||
X15202.1 | - | 6, 7 | 939 | |||
Y00769.1 | - | 7, 8 | 1034 |