Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 16 | 2276 | ||
NM_001164392.1 | NP_001157864.1 | 14 | 2185 | |||
NM_001164393.1 | NP_001157865.1 | 13 | 2075 | |||
NM_007358.3 | NP_031384.1 | 15 | 2356 | |||
XM_011541016.1 | XP_011539318.1 | 15 | 2312 | |||
XM_011541019.1 | XP_011539321.1 | 15 | 2104 | |||
XM_017000676.1 | XP_016856165.1 | 15 | 2211 | |||
XM_017000677.1 | XP_016856166.1 | 14 | 2141 | |||
XM_017000678.1 | XP_016856167.1 | 15 | 2094 | |||
XM_017000679.1 | XP_016856168.1 | 14 | 2188 | |||
GenBank mRNA | AJ010014.1 | - | 15 | 2311 |