Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014267.5 | NP_055082.1 | 4 | 441 | ||
XM_017017143.1 | XP_016872632.1 | 4 | 2700 | |||
GenBank mRNA | AK057169.1 | - | 2 | 1509 | ||
AK093998.1 | - | 1 | 1254 | |||
AK094549.1 | - | 1 | 2152 | |||
AK315382.1 | - | 4 | 395 | |||
BC007103.1 | - | 4 | 395 | 4281282 | ||
BC016352.1 | - | 4 | 399 | 4082845 | ||
BC020937.1 | - | 4 | 395 | 4281930 | ||
BC103747.1 | - | 4 | 422 | 6151644 | ||
BP210918.1 | - | 5 | 479 | |||
BX648095.1 | - | 1 | 3725 | |||
CR456882.1 | - | 4 | 263 | |||
U51678.1 | - | 4 | 298 |