Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 7 | 797 | ||
XM_005245817.1 | XP_005245874.1 | 6 | 668 | |||
XM_011541159.1 | XP_011539461.1 | 7 | 991 | |||
GenBank mRNA | AF081282.1 | - | 7 | 724 | ||
AK299180.1 | - | 6 | 668 | |||
AK311092.1 | - | 4 | 475 | |||
AK311355.1 | - | 5 | 542 | |||
AL136627.1 | - | 7 | 779 | |||
AY071927.1 | - | 7 | 774 | |||
AY358650.1 | - | 7 | 760 | |||
BC007341.2 | - | 7 | 745 | 3688672 | ||
BC066908.1 | - | 7 | 736 | 4817784 | ||
BX537683.1 | - | 1 | 3721 | |||
JX644905.1 | - | 7 | 797 | |||
JX644906.1 | - | 7 | 797 |