Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014857.4 | NP_055672.3 | 4 | 721 | ||
XM_005245680.1 | XP_005245737.1 | 4 | 651 | |||
XM_005245681.2 | XP_005245738.1 | 4 | 610 | |||
XM_006711693.1 | XP_006711756.1 | 4 | 651 | |||
XM_011510223.2 | XP_011508525.1 | 5 | 678 | |||
XM_017002994.1 | XP_016858483.1 | 4 | 608 | |||
GenBank mRNA | AB449904.1 | - | 3 | 444 | ||
AK095838.2 | - | 4 | 618 | |||
AL157958.1 | - | 4 | 545 | |||
BC041888.1 | - | 4 | 614 | 5298493 |