Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258210.1 | NP_001245139.1 | 2 | 1782 | ||
NM_001318477.1 | NP_001305406.1 | 2 | 2679 | |||
NM_001318478.1 | NP_001305407.1 | 2 | 1879 | |||
NM_001318479.1 | NP_001305408.1 | 2 | 1690 | |||
NM_015516.3 | NP_056331.2 | 2 | 1825 | |||
GenBank mRNA | AF191019.1 | - | 1 | 1662 | ||
AK075476.1 | - | 2 | 1737 | |||
AK091475.1 | - | 3 | 1642 | |||
AK126536.1 | - | 2 | 2679 | |||
AL110276.1 | - | 1 | 1321 | |||
AY358317.1 | - | 2 | 1716 | |||
BC020975.1 | - | 2 | 1703 | 3848274 |