Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 4 | 1071 | ||
NM_014955.2 | NP_055770.1 | 4 | 1214 | |||
NM_015935.4 | NP_057019.3 | 4 | 1539 | |||
XM_017001455.1 | XP_016856944.1 | 3 | 1033 | |||
XM_017001456.1 | XP_016856945.1 | 3 | 1033 | |||
XM_017001457.1 | XP_016856946.1 | 3 | 1033 | |||
XM_017001458.1 | XP_016856947.1 | 4 | 1656 | |||
XM_017001459.1 | XP_016856948.1 | 4 | 1206 | |||
GenBank mRNA | AB020666.2 | - | 4 | 1294 | ||
AB242174.1 | - | 4 | 1464 | |||
AB242175.1 | - | 4 | 1452 | |||
AF132936.1 | - | 4 | 1464 | |||
AK001172.1 | - | 2 | 226 | |||
AK027552.1 | - | 4 | 1458 | |||
AK027621.1 | - | 4 | 1153 | |||
AK027886.1 | - | 3 | 1010 | |||
AK129561.1 | - | 3 | 887 | |||
AK223493.1 | - | 4 | 1500 | |||
AK291740.1 | - | 4 | 1486 | |||
AK304471.1 | - | 5 | 1320 | |||
AL049669.1 | - | 4 | 996 | |||
BC006814.1 | - | 1 | 89 | |||
BC029083.1 | - | 4 | 1293 | 4509086 |