Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077394.1 | NP_001070862.1 | 8 | 1015 | ||
NM_001077395.1 | NP_001070863.1 | 8 | 994 | |||
NM_015958.2 | NP_057042.2 | 8 | 997 | |||
XM_005270938.1 | XP_005270995.1 | 7 | 893 | |||
XM_005270939.4 | XP_005270996.1 | 7 | 805 | |||
XM_005270941.3 | XP_005270998.1 | 7 | 780 | |||
XM_006710683.3 | XP_006710746.1 | 6 | 652 | |||
XM_011541569.1 | XP_011539871.1 | 7 | 890 | |||
XM_011541570.2 | XP_011539872.1 | 7 | 797 | |||
XM_017001460.1 | XP_016856949.1 | 8 | 1017 | |||
XM_017001461.1 | XP_016856950.1 | 7 | 804 | |||
XM_017001462.1 | XP_016856951.1 | 7 | 799 | |||
GenBank mRNA | AF132964.1 | - | 8 | 1002 | ||
AF161492.1 | - | 7 | 853 | |||
AF248965.1 | - | 8 | 966 | |||
AK125485.1 | - | 9 | 1366 | |||
AK225469.1 | - | 8 | 972 | |||
AK225707.1 | - | 8 | 972 | |||
AK225951.1 | - | 8 | 972 | |||
AK289351.1 | - | 8 | 983 | |||
AK298181.1 | - | 6 | 719 | |||
BC009620.2 | - | 2 | 257 | |||
BC053857.1 | - | 8 | 963 | 5503038 |