Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286229.1 | NP_001273158.1 | 8 | 992 | ||
NM_001286230.1 | NP_001273159.1 | 9 | 1096 | |||
NM_016448.3 | NP_057532.3 | 10 | 1222 | |||
XM_011509614.1 | XP_011507916.1 | 10 | 926 | |||
GenBank mRNA | AF195765.1 | - | 10 | 986 | ||
AF345896.1 | - | 10 | 1007 | |||
AK000742.1 | - | 10 | 1040 | |||
AK001261.1 | - | 1 | 29 | |||
AK027651.1 | - | 8 | 699 | |||
AK292343.1 | - | 10 | 1094 | |||
AK303346.1 | - | 9 | 899 | |||
BC033297.1 | - | 10 | 963 | 4342989 | ||
BC033540.1 | - | 10 | 1004 | 4826434 | ||
DQ641253.1 | - | 10 | 884 |