Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 5 | 1652 | ||
NM_001297749.1 | NP_001284678.1 | 4 | 1475 | |||
NM_001297750.1 | NP_001284679.1 | 7 | 1849 | |||
NM_001297751.1 | NP_001284680.1 | 6 | 1793 | |||
NM_016511.3 | NP_057595.2 | 6 | 1751 | |||
XM_011520687.2 | XP_011518989.1 | 7 | 1832 | |||
GenBank mRNA | AF200949.1 | - | 6 | 1689 | ||
AK075114.1 | - | 6 | 1671 | |||
AK091989.1 | - | 5 | 1603 | |||
AY358587.1 | - | 6 | 1751 | |||
BC039072.1 | - | 6 | 1657 | 5178017 | ||
BC067746.1 | - | 6 | 1671 | 30337675 |