Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017619.3 | NP_060089.1 | 3, 4 | 482 | ||
GenBank mRNA | AK057799.1 | - | 3, 4 | 461 | ||
AK289844.1 | - | 3, 4 | 747 | |||
AY099329.1 | - | 3, 4 | 467 | |||
BC010697.1 | - | 3, 4 | 458 | 3873751 |