Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018060.3 | NP_060530.3 | 4 | 773 | ||
GenBank mRNA | AK001188.1 | - | 2 | 248 | ||
AK291432.1 | - | 4 | 582 | |||
AY267462.1 | - | 4 | 784 | |||
AY952883.1 | - | 4 | 687 | |||
BC040376.1 | - | 4 | 506 | |||
BC047880.1 | - | 3 | 300 | |||
BC137438.1 | - | 4 | 743 | 9021436 | ||
D28500.1 | - | 4 | 614 |