Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018122.4 | NP_060592.2 | 9, 10 | 1404 | ||
XM_006711427.3 | XP_006711490.1 | 9, 10 | 1361 | |||
XM_011509711.2 | XP_011508013.1 | 9, 10 | 1361 | |||
GenBank mRNA | AK001376.1 | - | 5, 6 | 408 | ||
AK022754.1 | - | 5, 6 | 409 | |||
AK290873.1 | - | 9, 10 | 1086 | |||
AL833633.1 | - | 9, 10 | 1089 | |||
BC045173.2 | - | 9, 10 | 1383 | 5273243 |