Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 2 | 276 | ||
XM_017001745.1 | XP_016857234.1 | 2 | 203 | |||
GenBank mRNA | AF306694.1 | - | 2 | 175 | ||
AF306695.1 | - | 2 | 190 | |||
AK001599.1 | - | 2 | 185 | |||
AK294539.1 | - | 2 | 173 | |||
AK298378.1 | - | 2 | 162 | |||
AL109978.1 | - | 1 | 98 | |||
AL833841.1 | - | 2 | 150 | |||
AY336749.1 | - | 2 | 186 | |||
BC006086.1 | - | 2 | 172 | 3946414 | ||
BC008772.1 | - | 2 | 177 | |||
BC014145.2 | - | 2 | 184 | 4762069 | ||
DB461237.1 | - | 2 | 278 |