Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282564.1 | NP_001269493.1 | 6 | 964 | ||
NM_018202.5 | NP_060672.2 | 8 | 1645 | |||
XM_005245931.2 | XP_005245988.1 | 8 | 1606 | |||
XM_011541704.2 | XP_011540006.1 | 8 | 1679 | |||
GenBank mRNA | AK001609.1 | - | 8 | 1556 | ||
AY845015.1 | - | 8 | 1575 | |||
AY845035.1 | - | 6 | 832 | |||
AY845036.1 | - | 3 | 439 | |||
BC032427.1 | - | 8 | 1513 | 5246967 |