Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123107
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
17
1918
NM_001244714.1
NP_001231643.1
14
1483
NM_001278224.1
NP_001265153.1
10
1002
NM_018291.3
NP_060761.3
16
1846
XM_011541730.1
XP_011540032.1
17
1842
XM_011541731.1
XP_011540033.1
17
1776
XM_011541732.1
XP_011540034.1
17
1819
XM_011541733.1
XP_011540035.1
17
1737
XM_011541735.1
XP_011540037.1
16
1665
XM_011541736.2
XP_011540038.1
15
1683
XM_017001643.1
XP_016857132.1
17
1948
XM_017001644.1
XP_016857133.1
17
1741
XM_017001645.1
XP_016857134.1
16
1770
XM_017001646.1
XP_016857135.1
16
1747
XM_017001647.1
XP_016857136.1
16
1875
XM_017001648.1
XP_016857137.1
18
1761
XM_017001649.1
XP_016857138.1
16
1792
XM_017001650.1
XP_016857139.1
18
2038
XM_017001651.1
XP_016857140.1
18
1894
XM_017001652.1
XP_016857141.1
15
1720
XM_017001653.1
XP_016857142.1
15
1652
XM_017001654.1
XP_016857143.1
15
1638
XM_017001655.1
XP_016857144.1
15
1547
XM_017001656.1
XP_016857145.1
18
1954
XM_017001657.1
XP_016857146.1
17
1742
XM_017001658.1
XP_016857147.1
18
1885
XM_017001659.1
XP_016857148.1
16
1775
XM_017001660.1
XP_016857149.1
16
1678
XM_017001661.1
XP_016857150.1
16
1835
XM_017001662.1
XP_016857151.1
16
1520
XM_017001663.1
XP_016857152.1
16
1695
XM_017001664.1
XP_016857153.1
15
1572
XM_017001665.1
XP_016857154.1
15
1479
XM_017001666.1
XP_016857155.1
14
1561
XM_017001667.1
XP_016857156.1
14
1615
XM_017001668.1
XP_016857157.1
15
1443
XM_017001669.1
XP_016857158.1
15
1538
XM_017001670.1
XP_016857159.1
15
2311
XM_017001671.1
XP_016857160.1
15
1439
XM_017001672.1
XP_016857161.1
15
1703
XM_017001673.1
XP_016857162.1
15
1760
XM_017001674.1
XP_016857163.1
15
1619
XM_017001675.1
XP_016857164.1
15
1680
XM_017001676.1
XP_016857165.1
14
1668
XM_017001677.1
XP_016857166.1
13
4013
XM_017001678.1
XP_016857167.1
13
5288
GenBank mRNA
AK001848.1
-
10
876
AK054842.1
-
14
1474
AK315649.1
-
16
1731
BC000610.2
-
10
977
BC014947.1
-
16
1731
4871664
BX647626.1
-
17
1890
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0