Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040697.2 | NP_001035787.1 | 7 | 764 | ||
NM_001261382.1 | NP_001248311.1 | 6 | 698 | |||
NM_001261383.1 | NP_001248312.1 | 6 | 698 | |||
NM_001261384.1 | NP_001248313.1 | 5 | 515 | |||
NM_001261385.1 | NP_001248314.1 | 7 | 721 | |||
NM_001261386.1 | NP_001248315.1 | 5 | 464 | |||
NM_018314.4 | NP_060784.3 | 7 | 764 | |||
XM_006718255.2 | XP_006718318.1 | 6 | 593 | |||
XM_011520201.2 | XP_011518503.1 | 3 | 2595 | |||
XM_017017985.1 | XP_016873474.1 | 6 | 691 | |||
XM_017017986.1 | XP_016873475.1 | 6 | 571 | |||
GenBank mRNA | AF503350.1 | - | 7 | 724 | ||
AK001930.1 | - | 7 | 721 | |||
AK295501.1 | - | 5 | 443 | |||
AK296835.1 | - | 6 | 658 | |||
AK301537.1 | - | 6 | 640 | |||
AK314521.1 | - | 7 | 764 | |||
AK316191.1 | - | 6 | 571 | |||
BC064566.1 | - | 6 | 638 | 5576632 | ||
CD109744.1 | - | 5 | 463 |