Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018638.4 | NP_061108.2 | 3, 4 | 854 | ||
XM_017019580.1 | XP_016875069.1 | 3, 4 | 851 | |||
XM_017019581.1 | XP_016875070.1 | 3, 4 | 851 | |||
GenBank mRNA | AF207600.2 | - | 3, 4 | 920 | ||
AK074425.1 | - | 3, 4 | 799 | |||
AL137750.1 | - | 3, 4 | 803 | |||
BC066907.1 | - | 3, 4 | 854 | 5260109 |