Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136204.2 | NP_001129676.1 | 1, 2 | 333 | ||
NM_018715.3 | NP_061185.1 | 2, 3 | 502 | |||
GenBank mRNA | AB040903.1 | - | 2, 3 | 433 | ||
AJ421269.1 | - | 2, 3 | 519 | |||
BC042141.1 | - | 1, 2 | 298 | 5932549 | ||
BC053908.1 | - | 1, 2 | 331 | 6502476 | ||
BC142946.1 | - | 1, 2 | 89 | |||
CR991144.1 | - | 1, 2 | 333 | |||
DR000303.1 | - | 2, 3 | 508 |