Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038707.1 | NP_001033796.1 | 6 | 1567 | ||
NM_020239.3 | NP_064624.1 | 5 | 1431 | |||
XM_017001847.1 | XP_016857336.1 | 6 | 1511 | |||
GenBank mRNA | AF187845.2 | - | 5 | 1354 | ||
AF286592.1 | - | 4 | 1906 | |||
BC012796.2 | - | 4 | 1175 | 3924598 |