Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020348.2 | NP_065081.2 | 5 | 2356 | ||
XM_005269692.3 | XP_005269749.2 | 5 | 2356 | |||
XM_011539629.1 | XP_011537931.1 | 5 | 2356 | |||
XM_011539630.1 | XP_011537932.1 | 5 | 2356 | |||
XM_011539631.2 | XP_011537933.1 | 5 | 2356 | |||
GenBank mRNA | AF169226.1 | - | 5 | 1179 | ||
AK056194.1 | - | 4 | 459 | |||
AK296545.1 | - | 5 | 1524 | |||
AK299346.1 | - | 5 | 1513 | |||
AK308326.1 | - | 4 | 403 | |||
AK308618.1 | - | 5 | 2097 | |||
AK310486.1 | - | 4 | 611 | |||
BC098103.1 | - | 5 | 1130 | 40006972 | ||
BC098134.1 | - | 5 | 1130 | |||
BC098279.1 | - | 5 | 1130 | 40006977 | ||
BC098307.1 | - | 5 | 1130 | 40006978 | ||
JF267782.1 | - | 3 | 286 |