Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020362.4 | NP_065095.2 | 6 | 1216 | ||
XM_011541804.1 | XP_011540106.1 | 5 | 1141 | |||
GenBank mRNA | AF218024.1 | - | 6 | 1197 | ||
AF221595.1 | - | 6 | 1208 | |||
AF271784.1 | - | 6 | 1212 | |||
BC017208.2 | - | 6 | 1177 | 3943360 |