Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAMBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020799.3 | NP_065850.1 | 4 | 786 | ||
XM_006717928.2 | XP_006717991.1 | 5 | 887 | |||
XM_006717930.3 | XP_006717993.1 | 4 | 1374 | |||
XM_011539983.1 | XP_011538285.1 | 5 | 1294 | |||
XM_011539985.1 | XP_011538287.1 | 5 | 617 | |||
XM_017016453.1 | XP_016871942.1 | 5 | 1611 | |||
XM_017016454.1 | XP_016871943.1 | 6 | 1095 | |||
XM_017016455.1 | XP_016871944.1 | 7 | 1550 | |||
XM_017016456.1 | XP_016871945.1 | 5 | 1207 | |||
XM_017016457.1 | XP_016871946.1 | 4 | 1133 | |||
XM_017016458.1 | XP_016871947.1 | 2 | 311 | |||
GenBank mRNA | AB010120.1 | - | 3 | 314 | ||
AB037794.1 | - | 4 | 1107 | |||
AK056086.1 | - | 4 | 715 | |||
AK092946.1 | - | 3 | 915 | |||
BC010846.1 | - | 4 | 704 | 3898533 | ||
BX649096.1 | - | 5 | 9209 |