Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320238.1 | NP_001307167.1 | 13 | 3591 | ||
NM_001320239.1 | NP_001307168.1 | 13 | 3535 | |||
NM_001320240.1 | NP_001307169.1 | 17 | 4066 | |||
NM_020850.2 | NP_065901.1 | 14 | 3613 | |||
XM_005256070.2 | XP_005256127.1 | 14 | 3611 | |||
XM_017023507.1 | XP_016878996.1 | 11 | 3127 | |||
XM_017023509.1 | XP_016878998.1 | 12 | 3116 | |||
GenBank mRNA | AB040897.1 | - | 14 | 3502 | ||
AK055634.1 | - | 1 | 1031 | |||
AY337313.1 | - | 14 | 3613 | |||
BC099917.1 | - | 14 | 3519 | 30528481 |