Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022078.2 | NP_071361.2 | 1 | 377 | ||
GenBank mRNA | AK022517.1 | - | 1 | 372 | ||
AK057930.1 | - | 1 | 372 | |||
AK303183.1 | - | 1 | 374 | |||
AL833886.1 | - | 1 | 359 | |||
BC007767.2 | - | 1 | 353 | 4298740 | ||
CR936767.1 | - | 1 | 377 | |||
DA513389.1 | - | 1 | 377 |