Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303251.1 | NP_001290180.1 | 3 | 791 | ||
NM_022090.4 | NP_071373.2 | 2 | 717 | |||
GenBank mRNA | AK026754.1 | - | 1 | 523 | ||
BC032603.1 | - | 3 | 738 | 5535434 | ||
BC112128.1 | - | 1 | 259 | 8327596 | ||
BC112130.1 | - | 1 | 259 | 8327598 | ||
BM457021.1 | - | 2 | 671 | |||
BQ959342.1 | - | 2 | 654 |