Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198978.1 | NP_001185907.1 | 3 | 251 | ||
NM_001198979.1 | NP_001185908.1 | 3 | 335 | |||
NM_001198980.1 | NP_001185909.1 | 3 | 266 | |||
NM_022733.2 | NP_073570.1 | 3 | 693 | |||
XM_011541960.1 | XP_011540262.1 | 4 | 700 | |||
GenBank mRNA | AK298702.1 | - | 3 | 231 | ||
AK303975.1 | - | 3 | 239 | |||
AK308280.1 | - | 3 | 335 | |||
AK313105.1 | - | 3 | 640 | |||
AK316218.1 | - | 3 | 266 | |||
AL133206.1 | - | 3 | 207 | |||
AL137764.1 | - | 3 | 220 | |||
BC014512.1 | - | 1 | 32 | |||
BC021133.1 | - | 3 | 343 | 4855320 | ||
DC411665.1 | - | 3 | 251 |