Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022774.1 | NP_073611.1 | 3 | 1666 | ||
XM_005271125.3 | XP_005271182.1 | 4 | 1914 | |||
XM_005271126.3 | XP_005271183.1 | 3 | 1726 | |||
XM_005271128.3 | XP_005271185.1 | 2 | 1663 | |||
XM_011541970.2 | XP_011540272.1 | 2 | 1608 | |||
XM_017002099.1 | XP_016857588.1 | 4 | 2338 | |||
XM_017002100.1 | XP_016857589.1 | 4 | 1893 | |||
XM_017002101.1 | XP_016857590.1 | 4 | 1864 | |||
XM_017002102.1 | XP_016857591.1 | 3 | 2266 | |||
XM_017002103.1 | XP_016857592.1 | 3 | 1692 | |||
XM_017002104.1 | XP_016857593.1 | 2 | 1645 | |||
GenBank mRNA | AF086542.1 | - | 1 | 397 | ||
AK021507.1 | - | 2 | 1184 | |||
AK023245.1 | - | 2 | 2494 | |||
AK024797.1 | - | 3 | 1666 | |||
AK094092.1 | - | 3 | 2488 | |||
BC021969.2 | - | 4 | 1827 | 3952573 |