Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282143.1 | NP_001269072.1 | 10 | 3725 | ||
NM_001282144.1 | NP_001269073.1 | 10 | 4133 | |||
NM_001282358.1 | NP_001269287.1 | 10 | 3657 | |||
NM_024618.3 | NP_078894.2 | 10 | 3752 | |||
XM_005271669.1 | XP_005271726.1 | 10 | 3976 | |||
XM_005271670.1 | XP_005271727.1 | 9 | 3629 | |||
XM_005271671.1 | XP_005271728.1 | 9 | 3803 | |||
XM_005271672.1 | XP_005271729.1 | 9 | 3626 | |||
XM_006718904.1 | XP_006718967.1 | 10 | 3307 | |||
XM_011542980.1 | XP_011541282.1 | 10 | 3640 | |||
GenBank mRNA | AB094095.1 | - | 10 | 3736 | ||
AK025131.1 | - | 5 | 2379 | |||
AK225223.1 | - | 5 | 2379 | |||
AL049456.1 | - | 1 | 394 | |||
BC013199.1 | - | 6 | 2153 | |||
BC110890.1 | - | 10 | 4098 | 4937963 |