Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024763.4 | NP_079039.4 | 8 | 1225 | ||
NM_207014.2 | NP_996897.2 | 8 | 1225 | |||
XM_011542161.2 | XP_011540463.1 | 8 | 1178 | |||
XM_011542162.2 | XP_011540464.1 | 8 | 1178 | |||
XM_017002355.1 | XP_016857844.1 | 5 | 598 | |||
XM_017002356.1 | XP_016857845.1 | 6 | 842 | |||
XM_017002357.1 | XP_016857846.1 | 8 | 1178 | |||
GenBank mRNA | AK026782.1 | - | 4 | 467 | ||
AK127011.1 | - | 6 | 1746 | |||
AK292830.1 | - | 9 | 1178 | |||
BC032406.1 | - | 8 | 1124 | |||
BC126379.1 | - | 8 | 1200 | 8992095 | ||
BC126381.1 | - | 8 | 1200 | 8992097 | ||
BX648840.1 | - | 8 | 1226 |