Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300800.1 | NP_001287729.1 | 8 | 1592 | ||
NM_024771.3 | NP_079047.2 | 8 | 1521 | |||
XM_006718689.3 | XP_006718752.1 | 8 | 1418 | |||
XM_011545254.2 | XP_011543556.1 | 9 | 1595 | |||
XM_011545255.2 | XP_011543557.1 | 9 | 1505 | |||
GenBank mRNA | AK023910.1 | - | 8 | 1490 | ||
BC024160.2 | - | 1 | 778 | |||
BC041617.1 | - | 8 | 1450 | 5921480 | ||
BC052298.1 | - | 8 | 1450 | 6292760 | ||
BX647309.1 | - | 7 | 2547 | |||
JF263566.1 | - | 8 | 1490 |