Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024829.5 | NP_079105.4 | 3 | 517 | ||
GenBank mRNA | AI370813.1 | - | 3 | 366 | ||
AK026315.1 | - | 3 | 280 | |||
AK225458.1 | - | 3 | 280 | |||
AK290914.1 | - | 3 | 378 | |||
BC063561.1 | - | 3 | 381 | 4993580 | ||
BU859334.1 | - | 3 | 514 | |||
CB993231.1 | - | 3 | 464 |