Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297698.1 | NP_001284627.1 | 4 | 1013 | ||
NM_024865.3 | NP_079141.2 | 4 | 1061 | |||
XM_011520850.1 | XP_011519152.1 | 4 | 1058 | |||
XM_011520852.1 | XP_011519154.1 | 4 | 986 | |||
GenBank mRNA | AB093576.1 | - | 4 | 1058 | ||
AK022643.1 | - | 4 | 1058 | |||
AK290896.1 | - | 4 | 1027 | |||
AY230262.1 | - | 4 | 842 | |||
AY578089.1 | - | 4 | 794 | |||
BC069807.1 | - | 1 | 990 | |||
BC098275.1 | - | 1 | 990 | |||
BC099704.1 | - | 1 | 990 |
Gene Symbol: NANOGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097770.1 | - | 4 | 703 | ||
AY455283.1 | - | 4 | 703 | |||
KC713782.1 | - | 3 | 977 |