Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271895.1 | NP_001258824.1 | 6, 7 | 777 | ||
NM_025150.4 | NP_079426.2 | 6, 7 | 777 | |||
NR_073514.1 | 5, 6 | 652 | ||||
XM_006711555.1 | XP_006711618.1 | 6, 7 | 740 | |||
XM_006711556.1 | XP_006711619.1 | 6, 7 | 740 | |||
XM_017002394.1 | XP_016857883.1 | 6, 7 | 740 | |||
XM_017002395.1 | XP_016857884.1 | 6, 7 | 740 | |||
GenBank mRNA | AK222814.1 | - | 6, 7 | 718 | ||
AK225368.1 | - | 6, 7 | 697 | |||
AK298773.1 | - | 5, 6 | 605 | |||
AK315507.1 | - | 6, 7 | 723 | |||
BC000541.2 | - | 6, 7 | 724 | 3163506 | ||
BC007824.2 | - | 6, 7 | 665 | |||
BC009997.2 | - | 6, 7 | 697 | 4128881 |