Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 5 | 628 | ||
NM_001281749.1 | NP_001268678.1 | 4 | 504 | |||
NM_025164.4 | NP_079440.3 | 4 | 504 | |||
XM_005271481.2 | XP_005271538.1 | 4 | 542 | |||
XM_005271482.4 | XP_005271539.2 | 4 | 542 | |||
XM_005271484.3 | XP_005271541.2 | 4 | 542 | |||
XM_011542721.2 | XP_011541023.1 | 4 | 504 | |||
XM_011542722.1 | XP_011541024.1 | 4 | 542 | |||
XM_011542723.2 | XP_011541025.1 | 4 | 385 | |||
XM_011542724.2 | XP_011541026.1 | 3 | 239 | |||
XM_011542725.2 | XP_011541027.1 | 2 | 122 | |||
XM_011542726.1 | XP_011541028.1 | 4 | 542 | |||
XM_017017424.1 | XP_016872913.1 | 4 | 542 | |||
XM_017017425.1 | XP_016872914.1 | 4 | 580 | |||
XM_017017426.1 | XP_016872915.1 | 2 | 454 | |||
GenBank mRNA | BC121188.1 | - | 4 | 397 | ||
BC128510.1 | - | 4 | 366 | 40081639 | ||
BC128511.1 | - | 5 | 484 | 40081640 |