Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031725.5 | NP_001026895.2 | 2 | 887 | ||
NM_001320181.1 | NP_001307110.1 | 2 | 887 | |||
NM_001320182.1 | NP_001307111.1 | 1 | 655 | |||
XM_005245519.1 | XP_005245576.1 | 2 | 796 | |||
XM_005245521.2 | XP_005245578.1 | 2 | 796 | |||
XM_006711562.1 | XP_006711625.1 | 2 | 796 | |||
XM_011510035.2 | XP_011508337.1 | 2 | 820 | |||
XM_017002431.1 | XP_016857920.1 | 2 | 1575 | |||
XM_017002432.1 | XP_016857921.1 | 2 | 1060 | |||
GenBank mRNA | AK299219.1 | - | 2 | 758 | ||
AK307569.1 | - | 815 | ||||
AL136611.1 | - | 2 | 800 | |||
BC014183.2 | - | 2 | 820 | |||
BC041801.1 | - | 2 | 878 | 5302098 |