Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031935.2 | NP_114141.2 | 8 | 1342 | ||
XM_011510038.2 | XP_011508340.1 | 8 | 1359 | |||
XM_011510041.2 | XP_011508343.1 | 8 | 1367 | |||
GenBank mRNA | AF156100.1 | - | 8 | 1342 |