Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286373.1 | NP_001273302.1 | 2 | 437 | ||
NM_001286374.1 | NP_001273303.1 | 2 | 437 | |||
NM_032174.5 | NP_115550.2 | 2 | 437 | |||
XM_006711572.2 | XP_006711635.1 | 2 | 312 | |||
XM_011510057.2 | XP_011508359.1 | 2 | 200 | |||
XM_017002481.1 | XP_016857970.1 | 2 | 275 | |||
XM_017002482.1 | XP_016857971.1 | 2 | 305 | |||
GenBank mRNA | AK022832.1 | - | 2 | 316 | ||
AK027704.1 | - | 2 | 316 | |||
AK297831.1 | - | 2 | 292 | |||
AK297848.1 | - | 2 | 316 | |||
AL523459.3 | - | 2 | 294 | |||
BC014946.1 | - | 1 | 290 | 4872309 | ||
BX537951.1 | - | 2 | 314 | |||
DA160426.1 | - | 2 | 333 | |||
DA199234.1 | - | 2 | 337 |