Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014765.1 | NP_001014765.1 | 7 | 1301 | ||
NM_001304790.1 | NP_001291719.1 | 6 | 834 | |||
NM_001304791.1 | NP_001291720.1 | 6 | 1040 | |||
NM_033182.5 | NP_149438.2 | 6 | 929 | |||
NM_183412.2 | NP_904319.1 | 5 | 915 | |||
NM_183413.2 | NP_904320.1 | 5 | 804 | |||
NR_130909.1 | 5 | 670 | ||||
XM_005263535.1 | XP_005263592.1 | 6 | 871 | |||
XM_005263536.4 | XP_005263593.1 | 7 | 1265 | |||
XM_005263537.1 | XP_005263594.1 | 6 | 1011 | |||
XM_011542435.1 | XP_011540737.1 | 7 | 1333 | |||
GenBank mRNA | AK055344.1 | - | 5 | 915 | ||
AK293721.1 | - | 6 | 848 | |||
AK296025.1 | - | 5 | 650 | |||
AK301418.1 | - | 6 | 909 | |||
AK316284.1 | - | 5 | 1048 | |||
AK316347.1 | - | 6 | 886 | |||
AY040878.1 | - | 6 | 885 | |||
BC007832.2 | - | 5 | 751 | 4054414 | ||
BU164151.1 | - | 6 | 758 | |||
CR749368.1 | - | 7 | 1290 |