Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014765.1 | NP_001014765.1 | 7 | 2646 | ||
NM_001304790.1 | NP_001291719.1 | 6 | 2179 | |||
NM_001304791.1 | NP_001291720.1 | 6 | 2385 | |||
NM_033182.5 | NP_149438.2 | 6 | 2274 | |||
NM_183412.2 | NP_904319.1 | 5 | 2260 | |||
NM_183413.2 | NP_904320.1 | 5 | 2149 | |||
NR_130909.1 | 5 | 2015 | ||||
XM_006711043.2 | XP_006711106.1 | 7 | 900 | |||
XM_006711045.2 | XP_006711108.1 | 7 | 931 | |||
XM_017002844.1 | XP_016858333.1 | 6 | 804 | |||
GenBank mRNA | AY040878.1 | - | 6 | 2230 | ||
CR749368.1 | - | 7 | 2635 |