Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320333.1 | NP_001307262.1 | 7, 8 | 1439 | ||
NM_052939.3 | NP_443171.2 | 7, 8 | 1424 | |||
NR_135215.1 | 7, 8 | 1439 | ||||
NR_135216.1 | 7, 8 | 1344 | ||||
NR_135217.1 | 6, 7 | 1154 | ||||
XM_006711145.1 | XP_006711208.1 | 7, 8 | 1354 | |||
GenBank mRNA | AF416901.1 | - | 7, 8 | 1303 | ||
AF416902.1 | - | 6, 7 | 1018 | |||
AF416904.1 | - | 6, 7 | 1537 | |||
AF416905.1 | - | 7, 8 | 1208 | |||
AF459027.1 | - | 7, 8 | 1424 | |||
AK098122.1 | - | 7, 8 | 1354 | |||
AK301257.1 | - | 6, 7 | 1560 | |||
AL833391.1 | - | 1, 2 | 1974 | |||
AY043466.1 | - | 6, 7 | 1228 | |||
BC028933.1 | - | 7, 8 | 1305 | 4340939 |