Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256409.1 | NP_001243338.1 | 4, 5 | 875 | ||
NM_052940.4 | NP_443172.1 | 3, 4 | 841 | |||
XM_006710327.3 | XP_006710390.1 | 4, 5 | 697 | |||
XM_006710328.3 | XP_006710391.1 | 4, 5 | 1210 | |||
GenBank mRNA | AK074521.1 | - | 3, 4 | 814 | ||
AK075201.1 | - | 3, 4 | 803 | |||
AK090881.1 | - | 4, 5 | 1124 | |||
AK095767.1 | - | 4, 5 | 794 | |||
BC013101.1 | - | 3, 4 | 680 | 3918785 |