Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2ML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282424.2 | NP_001269353.1 | 23 | 2998 | ||
NM_144670.5 | NP_653271.2 | 34 | 4483 | |||
XM_011520566.2 | XP_011518868.1 | 34 | 4440 | |||
XM_011520567.2 | XP_011518869.1 | 34 | 4440 | |||
XM_017018868.1 | XP_016874357.1 | 34 | 4437 | |||
XM_017018869.1 | XP_016874358.1 | 34 | 4437 | |||
XM_017018870.1 | XP_016874359.1 | 34 | 4401 | |||
GenBank mRNA | AK057908.1 | - | 5 | 485 | ||
AK096448.1 | - | 12 | 1472 | |||
AK122624.1 | - | 28 | 3569 | |||
AK123591.1 | - | 17 | 2136 | |||
AK123592.1 | - | 23 | 2953 | |||
AK123601.1 | - | 31 | 3860 | |||
AK290355.1 | - | 34 | 4336 | |||
AK302555.1 | - | 23 | 2998 | |||
AL832139.1 | - | 34 | 4334 | |||
AL832750.1 | - | 12 | 1208 | |||
AL833479.1 | - | 32 | 3999 | |||
BC093840.1 | - | 5 | 436 | |||
BC112131.1 | - | 5 | 436 | |||
BX647674.1 | - | 16 | 1938 | |||
BX648231.1 | - | 35 | 4335 |