Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098510.1 | NP_001091980.1 | 3, 4 | 454 | ||
NM_001243135.1 | NP_001230064.1 | 3, 4 | 293 | |||
NM_001243136.1 | NP_001230065.1 | 2, 3 | 301 | |||
NM_145065.2 | NP_659502.2 | 4, 5 | 526 | |||
XM_011544884.2 | XP_011543186.1 | 4, 5 | 529 | |||
GenBank mRNA | AF487456.1 | - | 3, 4 | 362 | ||
AF487457.1 | - | 2, 3 | 290 | |||
AK074201.1 | - | 4, 5 | 464 | |||
AK094060.1 | - | 3, 4 | 293 | |||
AL834395.1 | - | 4, 5 | 473 | |||
BC021256.2 | - | 4, 5 | 467 | 3535985 | ||
BC025723.1 | - | 3, 4 | 375 | 5198999 | ||
BX341733.2 | - | 2, 3 | 238 |