Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114106.2 | NP_001107578.1 | 14 | 1948 | ||
NM_001258340.1 | NP_001245269.1 | 14 | 1945 | |||
NM_001258341.1 | NP_001245270.1 | 14 | 1849 | |||
NM_001258342.1 | NP_001245271.1 | 13 | 1840 | |||
NM_001258343.1 | NP_001245272.1 | 13 | 1744 | |||
NM_001301079.1 | NP_001288008.1 | 13 | 1787 | |||
NM_152369.4 | NP_689582.2 | 13 | 1883 | |||
XM_005270440.2 | XP_005270497.1 | 14 | 1843 | |||
XM_005270441.2 | XP_005270498.1 | 14 | 1885 | |||
XM_011540632.2 | XP_011538934.1 | 13 | 1812 | |||
XM_011540633.2 | XP_011538935.1 | 13 | 1650 | |||
XM_017000262.1 | XP_016855751.1 | 13 | 1822 | |||
XM_017000263.1 | XP_016855752.1 | 14 | 1882 | |||
GenBank mRNA | AK300193.1 | - | 13 | 1681 | ||
AK301288.1 | - | 14 | 1689 | |||
AK303906.1 | - | 13 | 1819 | |||
AK316123.1 | - | 16 | 1790 | |||
AY358659.1 | - | 10 | 1402 | |||
BC033858.1 | - | 14 | 1911 | |||
BC040448.1 | - | 1 | 97 | |||
BC053877.1 | - | 13 | 1806 | 5760394 | ||
BX648225.1 | - | 13 | 1811 |